Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HYDIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270974.2 | NP_001257903.1 | 35 | 5483 | ||
XM_006721206.2 | XP_006721269.1 | 35 | 5377 | |||
XM_011523146.2 | XP_011521448.1 | 36 | 5512 | |||
XM_011523147.1 | XP_011521449.1 | 36 | 5479 | |||
XM_011523148.1 | XP_011521450.1 | 36 | 5552 | |||
XM_011523151.1 | XP_011521453.1 | 35 | 5409 | |||
XM_017023346.1 | XP_016878835.1 | 35 | 5655 | |||
XM_017023347.1 | XP_016878836.1 | 23 | 3894 | |||
XM_017023348.1 | XP_016878837.1 | 23 | 3634 |
Gene Symbol: HYDIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103556.1 | 28 | 4440 | |||
GenBank mRNA | AB058767.1 | - | 2 | 172 | ||
AK027571.1 | - | 4 | 426 |