Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC554223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132323.1 | 3 | 726 | |||
GenBank mRNA | BC051362.1 | - | 3 | 726 | 4589959 |