Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003309.3 | NP_003300.1 | 1 | 3730 | ||
GenBank mRNA | AK096882.1 | - | 1 | 150 |