Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIPA1L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015073.2 | NP_055888.1 | 3 | 957 | ||
XM_005258671.4 | XP_005258728.1 | 3 | 1398 | |||
XM_011526657.2 | XP_011524959.1 | 3 | 1752 | |||
XM_017026517.1 | XP_016882006.1 | 4 | 1852 | |||
XM_017026518.1 | XP_016882007.1 | 3 | 1354 | |||
GenBank mRNA | AY168880.1 | - | 1 | 703 | ||
BC036863.1 | - | 3 | 929 | |||
BC150620.1 | - | 3 | 936 | 9056990 |