Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GNTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009905.2 | NP_001009905.1 | 6 | 481 | ||
NM_001320742.1 | NP_001307671.1 | 7 | 599 | |||
NM_001320743.1 | NP_001307672.1 | 2 | 1076 | |||
NR_135465.1 | 6 | 481 | ||||
NR_135466.1 | 2 | 1067 | ||||
XM_006722272.3 | XP_006722335.1 | 6 | 515 | |||
XM_011523532.2 | XP_011521834.1 | 5 | 449 | |||
XM_011523535.1 | XP_011521837.1 | 3 | 1418 | |||
XM_017024205.1 | XP_016879694.1 | 6 | 515 | |||
XM_017024206.1 | XP_016879695.1 | 3 | 202 | |||
GenBank mRNA | AK054757.1 | - | 2 | 1067 | ||
AY304503.1 | - | 6 | 479 | |||
AY634364.1 | - | 6 | 481 | |||
BC019623.1 | - | 4 | 275 |