Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLIC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114086.1 | NP_001107558.1 | 6 | 4671 | ||
NM_016929.4 | NP_058625.2 | 6 | 4435 | |||
NR_045672.1 | 3 | 4013 | ||||
NR_045673.1 | 5 | 4087 | ||||
XM_017010953.1 | XP_016866442.1 | 7 | 4916 | |||
GenBank mRNA | AJ420547.1 | - | 1 | 49 | ||
AK025350.1 | - | 1 | 776 | |||
AL049313.1 | - | 1 | 891 | |||
BC020923.1 | - | 1 | 199 | |||
DQ679794.1 | - | 6 | 4403 |