Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEFB114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037499.1 | NP_001032588.1 | 2 | 107 | ||
GenBank mRNA | AY122470.1 | - | 1 | 19 | ||
DQ012018.1 | - | 2 | 107 |