Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003468.3 | NP_003459.2 | 2 | 5030 | ||
GenBank mRNA | AK000125.1 | - | 1 | 233 | ||
AK024850.1 | - | 1 | 941 | |||
AL136852.1 | - | 1 | 573 | |||
BC095474.1 | - | 1 | 835 | |||
CR533442.1 | - | 1 | 206 |
Set Membership: |
G-protein coupled receptor |