Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TGFBRAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142621.2 | NP_001136093.1 | 12 | 4525 | ||
NM_004257.5 | NP_004248.2 | 12 | 4528 | |||
XM_017005339.1 | XP_016860828.1 | 11 | 4021 | |||
GenBank mRNA | BC019841.1 | - | 1 | 689 |