Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080467.2 | NP_001073936.1 | 38, 39 | 5602 | ||
GenBank mRNA | AB032945.2 | - | 38, 39 | 5332 | ||
AB290160.1 | - | 38, 39 | 5422 | |||
AK025336.1 | - | 17, 18 | 2667 | |||
AK093149.1 | - | 16, 17 | 2282 | |||
AK301976.1 | - | 10, 11 | 1753 | |||
AL133643.1 | - | 16, 17 | 2301 | |||
AY274809.1 | - | 16, 17 | 2281 | |||
BC033527.1 | - | 7, 8 | 1240 |