Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NELFB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015456.4 | NP_056271.3 | 2, 3 | 479 | ||
GenBank mRNA | AF464935.1 | - | 2, 3 | 284 | ||
AK091056.1 | - | 2, 3 | 472 | |||
BC011892.1 | - | 1, 2 | 188 | 3941719 | ||
BC019029.1 | - | 2, 3 | 438 | |||
DA210427.1 | - | 2, 3 | 473 |