Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015310.3 | NP_056125.3 | 16 | 10235 | ||
NM_206909.2 | NP_996792.1 | 13 | 8639 | |||
XM_017013258.1 | XP_016868747.1 | 16 | 11544 | |||
XM_017013259.1 | XP_016868748.1 | 16 | 11522 | |||
XM_017013260.1 | XP_016868749.1 | 16 | 10295 | |||
XM_017013261.1 | XP_016868750.1 | 15 | 10509 | |||
XM_017013265.1 | XP_016868754.1 | 13 | 9045 | |||
GenBank mRNA | AK313826.1 | - | 1 | 496 | ||
AL353936.1 | - | 1 | 1420 | |||
BC011238.1 | - | 1 | 512 |