Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AFMID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010982.4 | NP_001010982.2 | 6, 7 | 529 | ||
NM_001145526.2 | NP_001138998.1 | 6, 7 | 529 | |||
XM_011524326.2 | XP_011522628.1 | 5, 6 | 461 | |||
XM_011524327.2 | XP_011522629.1 | 7, 8 | 697 | |||
XM_011524328.2 | XP_011522630.1 | 7, 8 | 669 | |||
XM_011524329.2 | XP_011522631.1 | 7, 8 | 711 | |||
XM_011524330.2 | XP_011522632.1 | 7, 8 | 715 | |||
XM_011524331.2 | XP_011522633.1 | 6, 7 | 611 | |||
XM_011524332.2 | XP_011522634.1 | 6, 7 | 583 | |||
XM_011524333.2 | XP_011522635.1 | 6, 7 | 488 | |||
GenBank mRNA | BC132824.1 | - | 6, 7 | 529 | 40146846 | |
BX648442.1 | - | 6, 7 | 485 |