Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IBTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300906.1 | NP_001287835.1 | 26 | 4204 | ||
NM_015525.3 | NP_056340.2 | 26 | 4249 | |||
XM_006715453.2 | XP_006715516.1 | 26 | 4789 | |||
XM_006715454.2 | XP_006715517.1 | 26 | 4744 | |||
GenBank mRNA | AB037838.2 | - | 25 | 3973 | ||
AF235049.1 | - | 3 | 668 | |||
AJ420588.1 | - | 1 | 133 | |||
AJ420593.1 | - | 2 | 153 | |||
AK126777.1 | - | 10 | 2114 | |||
AL050018.1 | - | 6 | 789 | |||
BC027490.1 | - | 4 | 433 | |||
BC038244.2 | - | 9 | 1087 | |||
BC042171.1 | - | 5 | 632 | |||
BC113696.1 | - | 25 | 3778 | 8322748 | ||
BC113698.1 | - | 25 | 3778 | 8322750 | ||
BC143736.1 | - | 24 | 3175 | 9052250 | ||
BC143745.1 | - | 25 | 3734 | 9052259 | ||
BC143748.1 | - | 23 | 3745 | |||
DQ005633.1 | - | 26 | 4249 | |||
DQ005635.1 | - | 2 | 779 |