Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYO7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080527.1 | NP_001073996.1 | 14 | 1692 | ||
XM_006712539.3 | XP_006712602.1 | 14 | 1805 | |||
XM_011511218.2 | XP_011509520.1 | 14 | 1805 | |||
XM_017004169.1 | XP_016859658.1 | 14 | 1805 |