Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEPRO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319109.1 | NP_001306038.1 | 7 | 1996 | ||
NM_001319110.1 | NP_001306039.1 | 6 | 1841 | |||
NM_001319111.1 | NP_001306040.1 | 8 | 1810 | |||
NM_001319112.1 | NP_001306041.1 | 8 | 2009 | |||
NM_001319114.1 | NP_001306043.1 | 8 | 2055 | |||
NM_001319115.1 | NP_001306044.1 | 8 | 2051 | |||
NM_015412.3 | NP_056227.2 | 9 | 2181 | |||
NR_134969.1 | 6 | 1934 | ||||
XM_005247276.2 | XP_005247333.1 | 9 | 2148 | |||
XM_011512635.2 | XP_011510937.1 | 9 | 1903 | |||
XM_011512636.2 | XP_011510938.1 | 9 | 1907 | |||
XM_011512638.2 | XP_011510940.1 | 8 | 1802 | |||
XM_017006102.1 | XP_016861591.1 | 9 | 2170 | |||
GenBank mRNA | AK024325.1 | - | 9 | 2139 | ||
AK301161.1 | - | 8 | 1969 | |||
AL117573.1 | - | 9 | 2120 | |||
BC014673.2 | - | 8 | 2639 | |||
BC067743.1 | - | 9 | 2141 | 4812681 | ||
CR749341.1 | - | 7 | 1997 |