Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YIF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039671.2 | NP_001034760.1 | 8 | 1520 | ||
NM_001039672.2 | NP_001034761.1 | 8 | 1538 | |||
NM_001039673.2 | NP_001034762.1 | 8 | 1529 | |||
NM_001145461.1 | NP_001138933.1 | 8 | 1514 | |||
NM_001145462.1 | NP_001138934.1 | 8 | 1490 | |||
NM_001145463.1 | NP_001138935.1 | 9 | 1606 | |||
GenBank mRNA | AK027130.1 | - | 1 | 213 | ||
BC025257.1 | - | 1 | 149 |