Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018941.3 | NP_061764.2 | 3 | 6912 | ||
GenBank mRNA | AK096736.1 | - | 1 | 1606 |