Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARFGEF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020340.4 | NP_065073.3 | 34 | 12082 | ||
GenBank mRNA | AB252196.1 | - | 34 | 12082 | ||
AK056626.1 | - | 1 | 1680 | |||
BC022980.1 | - | 1 | 600 |