Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC31B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015490.3 | NP_056305.1 | 11 | 1440 | ||
GenBank mRNA | AF274863.1 | - | 11 | 1436 | ||
AK128343.1 | - | 5 | 1574 | |||
AL080141.1 | - | 4 | 548 | |||
BC034946.1 | - | 10 | 1946 | |||
BC044569.1 | - | 10 | 1939 | |||
BC053949.1 | - | 10 | 1939 | |||
BC071580.1 | - | 9 | 1809 | |||
BC144313.1 | - | 10 | 1378 | 9052833 | ||
BX640914.1 | - | 10 | 1287 |