Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015557.2 | NP_056372.1 | 15, 16 | 2540 | ||
GenBank mRNA | AF425231.1 | - | 15, 16 | 2529 | ||
AK091386.1 | - | 6, 7 | 1016 | |||
AK127046.1 | - | 15, 16 | 2429 | |||
AL110281.1 | - | 4, 5 | 577 | |||
AL117491.1 | - | 5, 6 | 733 | |||
AL137693.1 | - | 4, 5 | 556 |