Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SENP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100409.2 | NP_001093879.1 | 8 | 1965 | ||
NM_001304792.1 | NP_001291721.1 | 8 | 1965 | |||
NM_015571.3 | NP_056386.2 | 9 | 1986 | |||
XM_006715457.3 | XP_006715520.1 | 10 | 1740 | |||
XM_011535746.2 | XP_011534048.1 | 8 | 1538 | |||
XM_011535747.2 | XP_011534049.1 | 8 | 1889 | |||
XM_017010719.1 | XP_016866208.1 | 9 | 1628 | |||
XM_017010720.1 | XP_016866209.1 | 9 | 944 | |||
XM_017010721.1 | XP_016866210.1 | 9 | 952 | |||
XM_017010722.1 | XP_016866211.1 | 10 | 1759 | |||
XM_017010723.1 | XP_016866212.1 | 10 | 1736 | |||
XM_017010724.1 | XP_016866213.1 | 10 | 2111 | |||
XM_017010725.1 | XP_016866214.1 | 7 | 1871 | |||
GenBank mRNA | AB018340.2 | - | 9 | 1014 | ||
AF196304.1 | - | 9 | 970 | |||
AF306508.1 | - | 9 | 985 | |||
AF307849.1 | - | 9 | 1045 | |||
AK001406.1 | - | 6 | 640 | |||
AK096455.1 | - | 8 | 1547 | |||
AK126921.1 | - | 8 | 1965 | |||
BC028583.1 | - | 8 | 1172 | 4838569 |