Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFRSF9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001561.5 | NP_001552.2 | 3 | 344 | ||
XM_006710618.3 | XP_006710681.1 | 2 | 991 | |||
XM_011541386.2 | XP_011539688.1 | 2 | 359 | |||
GenBank mRNA | BC006196.2 | - | 2 | 204 | 2924109 | |
L12964.2 | - | 2 | 222 | |||
U03397.1 | - | 2 | 202 |