Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017439.3 | NP_059135.2 | 15 | 1155 | ||
XM_011516325.2 | XP_011514627.1 | 15 | 1168 | |||
XM_011516326.2 | XP_011514628.1 | 15 | 1168 | |||
XM_011516327.2 | XP_011514629.1 | 15 | 1168 | |||
XM_011516330.2 | XP_011514632.1 | 8 | 614 | |||
XM_011516331.2 | XP_011514633.1 | 15 | 1168 | |||
XM_011516332.2 | XP_011514634.1 | 15 | 1168 | |||
XM_017012343.1 | XP_016867832.1 | 15 | 1168 | |||
XM_017012344.1 | XP_016867833.1 | 15 | 1176 | |||
XM_017012345.1 | XP_016867834.1 | 15 | 1168 | |||
XM_017012346.1 | XP_016867835.1 | 13 | 1037 | |||
XM_017012347.1 | XP_016867836.1 | 13 | 1050 | |||
XM_017012348.1 | XP_016867837.1 | 16 | 1244 | |||
XM_017012349.1 | XP_016867838.1 | 15 | 1615 | |||
XM_017012350.1 | XP_016867839.1 | 15 | 1168 | |||
XM_017012351.1 | XP_016867840.1 | 15 | 1168 | |||
GenBank mRNA | AK026747.1 | - | 1 | 39 | ||
AL834358.1 | - | 14 | 972 | |||
BC110797.1 | - | 15 | 1155 |