Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3YL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159597.2 | NP_001153069.1 | 7, 8 | 813 | ||
NM_001282682.1 | NP_001269611.1 | 9, 10 | 918 | |||
NM_001282687.1 | NP_001269616.1 | 9, 10 | 967 | |||
NM_015677.3 | NP_056492.2 | 7, 8 | 813 | |||
NR_104223.1 | 9, 10 | 2372 | ||||
NR_104225.1 | 7, 8 | 1008 | ||||
NR_104226.1 | 9, 10 | 1533 | ||||
GenBank mRNA | AK026507.1 | - | 7, 8 | 748 | ||
AK026586.1 | - | 7, 8 | 1008 | |||
AK123829.1 | - | 10, 11 | 2611 | |||
AK292312.1 | - | 9, 10 | 862 | |||
AL050373.1 | - | 9, 10 | 980 | |||
BC008374.1 | - | 7, 8 | 722 | 3996066 | ||
BC008375.1 | - | 7, 8 | 740 | 3996120 | ||
BC043234.1 | - | 9, 10 | 2377 | |||
CR457361.1 | - | 6, 7 | 700 |