Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024356.2 | 1 | 595 | |||
GenBank mRNA | BX115749.1 | - | 1 | 320 | ||
CK906195.1 | - | 1 | 100 |