Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATP5S | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003803.2 | NP_001003803.1 | 2, 3 | 853 | ||
NM_001003805.2 | NP_001003805.1 | 2, 3 | 853 | |||
NM_015684.3 | NP_056499.2 | 2, 3 | 853 | |||
NR_033761.1 | 3, 4 | 889 | ||||
XM_005267537.3 | XP_005267594.1 | 2, 3 | 760 | |||
XM_011536649.2 | XP_011534951.1 | 2, 3 | 742 | |||
XM_011536650.2 | XP_011534952.1 | 2, 3 | 744 | |||
XM_011536651.2 | XP_011534953.1 | 2, 3 | 744 | |||
XM_011536652.1 | XP_011534954.1 | 2, 3 | 142 | |||
XM_011536653.2 | XP_011534955.1 | 3, 4 | 184 | |||
XM_011536654.2 | XP_011534956.1 | 2, 3 | 748 | |||
XM_011536655.2 | XP_011534957.1 | 2, 3 | 748 | |||
XM_011536656.2 | XP_011534958.1 | 2, 3 | 780 | |||
XM_011536657.2 | XP_011534959.1 | 2, 3 | 291 | |||
XM_017021220.1 | XP_016876709.1 | 2, 3 | 224 | |||
GenBank mRNA | AF052186.1 | - | 2, 3 | 805 | ||
AK290008.1 | - | 2, 3 | 541 | |||
AY052377.1 | - | 2, 3 | 276 | |||
BC011549.1 | - | 2, 3 | 853 | 4554461 | ||
BC087851.1 | - | 3, 4 | 658 | |||
BG699167.1 | - | 2, 3 | 189 | |||
BU189283.1 | - | 2, 3 | 164 | |||
U79253.1 | - | 2, 3 | 804 |