Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015703.4 | NP_056518.2 | 3 | 348 | ||
GenBank mRNA | AK092492.1 | - | 1 | 874 | ||
AY039240.1 | - | 2 | 220 | |||
BC031838.1 | - | 2 | 263 | |||
BC035992.1 | - | 3 | 327 | |||
BC041639.1 | - | 3 | 306 | |||
BC042335.1 | - | 3 | 309 | |||
BC073834.1 | - | 3 | 321 | 5533064 | ||
BC121118.1 | - | 3 | 331 | 40120513 | ||
BC121119.1 | - | 3 | 331 | 40120514 | ||
BP314581.1 | - | 3 | 348 |