Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014911.3 | NP_055726.3 | 22 | 17733 | ||
GenBank mRNA | BX640963.1 | - | 2 | 1412 |