Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MANEAL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001031740.2 | NP_001026910.1 | 3 | 793 | ||
NM_001113482.1 | NP_001106954.1 | 3 | 793 | |||
GenBank mRNA | AB188490.1 | - | 3 | 1012 | ||
AK301399.1 | - | 3 | 387 | |||
BC063587.1 | - | 3 | 250 | |||
BC077730.1 | - | 3 | 793 | 6265730 |