Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAPKAPK5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017020127.1 | XP_016875616.1 | 15 | 7974 | ||
XM_017020128.1 | XP_016875617.1 | 15 | 6425 | |||
XM_017020132.1 | XP_016875621.1 | 16 | 8022 | |||
XM_017020133.1 | XP_016875622.1 | 15 | 7794 | |||
XM_017020134.1 | XP_016875623.1 | 15 | 7620 | |||
XM_017020135.1 | XP_016875624.1 | 14 | 7703 | |||
XM_017020136.1 | XP_016875625.1 | 15 | 7788 | |||
XM_017020138.1 | XP_016875627.1 | 11 | 7181 |
Gene Symbol: ADAM1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036636.1 | 1 | 1779 |