Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242699.1 | NP_001229628.1 | 10 | 1358 | ||
XM_006717835.3 | XP_006717898.1 | 10 | 2640 | |||
XM_006717836.3 | XP_006717899.1 | 9 | 1175 | |||
XM_017016241.1 | XP_016871730.1 | 10 | 2640 |