Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242699.1 | NP_001229628.1 | 14 | 2208 | ||
XM_006717836.3 | XP_006717899.1 | 13 | 2025 | |||
XM_011539797.2 | XP_011538099.1 | 13 | 3364 | |||
XM_011539799.2 | XP_011538101.1 | 11 | 4696 | |||
XM_017016241.1 | XP_016871730.1 | 12 | 4822 | |||
GenBank mRNA | BX647301.1 | - | 7 | 1257 |