Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007011.7 | NP_008942.3 | 15 | 7308 | ||
NM_152924.4 | NP_690888.1 | 11 | 6910 | |||
XM_005254832.1 | XP_005254889.1 | 14 | 6946 | |||
XM_005254833.1 | XP_005254890.1 | 11 | 6554 | |||
XM_011521165.1 | XP_011519467.1 | 12 | 6759 | |||
GenBank mRNA | AK095685.1 | - | 1 | 673 | ||
AK125089.1 | - | 1 | 2543 | |||
AK127559.1 | - | 1 | 3301 | |||
BC017905.1 | - | 1 | 799 | |||
CR749694.1 | - | 1 | 2156 |