Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030641.3 | NP_085144.1 | 3 | 8342 | ||
XM_011530392.2 | XP_011528694.1 | 4 | 8519 | |||
GenBank mRNA | AK097266.1 | - | 1 | 454 |