Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242672.1 | NP_001229601.1 | 4 | 675 | ||
XM_016999990.1 | XP_016855479.1 | 4 | 747 |