Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC5AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304359.1 | NP_001291288.1 | 16660 | |||
GenBank mRNA | AJ001402.1 | - | 18 | 2894 | ||
AL833060.1 | - | 12 | 1036 | |||
BC033831.1 | - | 13 | 1374 | |||
U06711.1 | - | 18 | 2769 | |||
Z48314.1 | - | 18 | 2503 |