Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF077202.1 | - | 2, 3 | 118 | ||
AF161448.1 | - | 1, 2 | 67 | |||
AK126594.1 | - | 2, 3 | 80 | |||
AY508979.1 | - | 1 | 101 | |||
AY794023.1 | - | 2, 3 | 102 | |||
BC001102.2 | - | 1 | 90 | |||
BC080531.1 | - | 2, 3 | 103 | 4875991 | ||
BU536461.1 | - | 2, 3 | 147 |