Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMDHD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145815.1 | NP_001139287.1 | 3, 4 | 453 | ||
NM_015944.3 | NP_057028.2 | 3, 4 | 453 | |||
XM_017023262.1 | XP_016878751.1 | 3, 4 | 464 | |||
XM_017023263.1 | XP_016878752.1 | 3, 4 | 464 | |||
XM_017023264.1 | XP_016878753.1 | 3, 4 | 464 | |||
XM_017023265.1 | XP_016878754.1 | 3, 4 | 464 | |||
XM_017023266.1 | XP_016878755.1 | 3, 4 | 412 | |||
XM_017023267.1 | XP_016878756.1 | 2, 3 | 316 | |||
GenBank mRNA | AK296877.1 | - | 3, 4 | 425 | ||
AK300747.1 | - | 2, 3 | 269 | |||
BC018734.1 | - | 3, 4 | 389 | 4869721 | ||
CN362870.1 | - | 2, 3 | 323 |