Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIRREL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286349.1 | NP_001273278.1 | 13 | 3940 | ||
NM_018240.6 | NP_060710.3 | 15 | 4240 | |||
XM_005245305.4 | XP_005245362.1 | 15 | 3945 | |||
GenBank mRNA | AK090554.1 | - | 1 | 623 |