Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GSS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000178.3 | NP_000169.1 | 7, 8 | 796 | ||
NM_001322494.1 | NP_001309423.1 | 7, 8 | 863 | |||
NM_001322495.1 | NP_001309424.1 | 7, 8 | 1625 | |||
GenBank mRNA | AB459500.1 | - | 3, 4 | 364 | ||
AK293144.1 | - | 7, 8 | 687 | |||
AK298292.1 | - | 5, 6 | 974 | |||
AK311421.1 | - | 5, 6 | 510 | |||
AK311567.1 | - | 5, 6 | 881 | |||
AK312492.1 | - | 7, 8 | 776 | |||
BC007927.1 | - | 7, 8 | 783 | 3946317 | ||
EU668322.1 | - | 7, 8 | 783 | |||
EU794618.1 | - | 7, 8 | 783 | |||
KU178105.1 | - | 6, 7 | 697 | |||
KU178106.1 | - | 4, 5 | 440 | |||
U34683.1 | - | 7, 8 | 737 |