Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POU5F1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480844.1 | - | 1 | 848 |
Gene Symbol: POU5F1P3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | GU480891.1 | - | 1 | 847 | ||
GU480897.1 | - | 1 | 846 |