Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYH7B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020884.4 | NP_065935.3 | 24 | 2670 | ||
XM_006723840.3 | XP_006723903.1 | 22 | 2311 | |||
XM_011528941.2 | XP_011527243.1 | 22 | 2335 | |||
XM_011528947.2 | XP_011527249.1 | 16 | 2708 | |||
XM_011528948.2 | XP_011527250.1 | 16 | 1867 | |||
XM_017027986.1 | XP_016883475.1 | 23 | 2371 | |||
XM_017027987.1 | XP_016883476.1 | 18 | 1886 |