Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIDT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040455.1 | NP_001035545.1 | 2, 3 | 350 | ||
GenBank mRNA | AF151799.1 | - | 2, 3 | 392 | ||
AK292501.1 | - | 2, 3 | 830 | |||
AK292974.1 | - | 2, 3 | 341 | |||
AK303023.1 | - | 2, 3 | 306 | |||
AY358442.1 | - | 2, 3 | 359 | |||
BC114522.1 | - | 2, 3 | 379 | 40035836 | ||
KC109742.1 | - | 2, 3 | 306 |