Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204856.1 | NP_001191785.1 | 7 | 1478 | ||
NM_024041.3 | NP_076946.1 | 7 | 1429 | |||
NR_037937.1 | 7 | 1379 | ||||
GenBank mRNA | AK056322.1 | - | 7 | 1394 |
Gene Symbol: TNFAIP8L2-SCNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204848.1 | NP_001191777.1 | 7 | 1341 |