Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008397.3 | NP_001008398.2 | 3 | 2489 | ||
NM_001306197.1 | NP_001293126.1 | 3 | 2336 | |||
NM_001306198.1 | NP_001293127.1 | 2 | 2227 | |||
NM_001306201.1 | NP_001293130.1 | 3 | 2210 | |||
NR_131336.1 | 4 | 2403 | ||||
NR_131337.1 | 4 | 2345 | ||||
NR_131338.1 | 3 | 2141 | ||||
NR_131340.1 | 3 | 2479 | ||||
XM_006714631.2 | XP_006714694.1 | 3 | 2455 | |||
GenBank mRNA | AK022110.1 | - | 1 | 504 | ||
BX537894.1 | - | 4 | 2353 | |||
BX647770.1 | - | 2 | 2177 |