Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146339.1 | NP_001139811.1 | 4 | 320 | ||
XM_005258856.3 | XP_005258913.1 | 2 | 243 | |||
XM_011526902.2 | XP_011525204.1 | 5 | 940 | |||
XM_011526903.2 | XP_011525205.1 | 4 | 910 | |||
XM_011526904.2 | XP_011525206.1 | 4 | 465 | |||
XM_011526905.2 | XP_011525207.1 | 3 | 315 |