Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JAK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000215.3 | NP_000206.2 | 14, 15 | 2015 | ||
XM_005259896.3 | XP_005259953.1 | 14, 15 | 2054 | |||
XM_011527990.1 | XP_011526292.1 | 14, 15 | 2054 | |||
GenBank mRNA | U09607.1 | - | 13, 14 | 2010 | ||
U31601.1 | - | 14, 15 | 2010 |