Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098416.3 | NP_001091886.1 | 15 | 2037 | ||
NM_001308090.1 | NP_001295019.1 | 15 | 2097 | |||
NM_015401.4 | NP_056216.2 | 16 | 2148 | |||
XM_011538478.1 | XP_011536780.1 | 16 | 2100 | |||
XM_011538479.1 | XP_011536781.1 | 17 | 2542 | |||
XM_011538480.1 | XP_011536782.1 | 15 | 1989 | |||
XM_011538481.1 | XP_011536783.1 | 16 | 1993 | |||
XM_011538482.1 | XP_011536784.1 | 16 | 2077 | |||
XM_017019455.1 | XP_016874944.1 | 15 | 2049 | |||
GenBank mRNA | AK024469.1 | - | 11 | 2128 | ||
AK122588.1 | - | 3 | 1768 | |||
AK128383.1 | - | 8 | 991 | |||
AK299292.1 | - | 16 | 2063 | |||
AK301545.1 | - | 16 | 2011 | |||
AK303481.1 | - | 15 | 2003 | |||
AL117455.1 | - | 12 | 1615 | |||
AY302468.1 | - | 16 | 1953 | |||
AY321367.1 | - | 15 | 1922 | |||
BC006453.1 | - | 5 | 572 | |||
BC020505.2 | - | 8 | 1115 | |||
BC064840.1 | - | 15 | 1931 | 6179239 | ||
BT009771.1 | - | 6 | 822 |