Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002536.2 | NP_002527.1 | 6 | 2218 | ||
GenBank mRNA | AK293521.1 | - | 5 | 2048 | ||
AK293875.1 | - | 6 | 2177 | |||
AK294776.1 | - | 6 | 2116 | |||
AK297924.1 | - | 3 | 1623 | |||
AK299561.1 | - | 1 | 1146 | |||
AK316199.1 | - | 4 | 1898 | |||
BC026050.1 | - | 1 | 1141 | |||
BC036942.1 | - | 3 | 1647 | |||
BC052609.1 | - | 5 | 2008 | |||
BC082255.1 | - | 4 | 2118 | |||
BC101817.1 | - | 6 | 2144 | 8069323 | ||
BC101819.1 | - | 6 | 2144 | 8069325 | ||
BC125088.1 | - | 6 | 2133 | 40117866 | ||
BC125089.1 | - | 5 | 1978 | |||
BG328213.1 | - | 1 | 328 | |||
BI088539.1 | - | 1 | 253 | |||
L08240.1 | - | 5 | 1997 |